Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191652.1 | NP_001178581.1 | 4 - 5 | 746 | 144 | |
XM_006247574.2 | XP_006247636.1 | 4 - 5 | 2089 | 144 | ||
XM_006247576.2 | XP_006247638.1 | 4 - 5 | 642 | 144 | ||
XM_008768349.1 | XP_008766571.1 | 4 - 5 | 704 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |