Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105809.2 | NP_001099279.1 | 5 - 6 | 568 | 83 | |
GenBank mRNA | BC166808.1 | - | 5 - 6 | 568 | 7124384 | 83 |
FQ228687.1 | - | 5 - 6 | 584 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |