Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stra13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750986.3 | XP_003751034.2 | 3 - 4 | 282 | 74 | |
XM_008768490.1 | XP_008766712.1 | 2 - 3 | 381 | 74 | ||
XM_008768491.1 | XP_008766713.1 | 3 - 4 | 459 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |