Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024269.1 | NP_001019440.1 | 5 - 6 | 854 | 90 | |
XM_006246006.2 | XP_006246068.1 | 6 - 7 | 819 | 90 | ||
XM_006246007.1 | XP_006246069.1 | 6 - 7 | 2223 | 90 | ||
XM_008767588.1 | XP_008765810.1 | 6 - 7 | 1682 | 90 | ||
GenBank mRNA | BC089960.1 | - | 5 - 6 | 854 | 7313914 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |