Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad51c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129777.1 | NP_001123249.1 | 3 - 4 | 568 | 123 | |
XM_006247106.1 | XP_006247168.1 | 3 - 4 | 646 | 123 | ||
GenBank mRNA | BC166862.1 | - | 3 - 4 | 475 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |