Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013859.1 | NP_001013881.1 | 3 - 4 | 383 | 67 | |
GenBank mRNA | BC083572.1 | - | 3 - 4 | 383 | 7112698 | 67 |
FQ228522.1 | - | 3 - 4 | 396 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |