Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147144.1 | NP_671485.1 | 4 - 5 | 442 | 123 | |
GenBank mRNA | AF525467.1 | - | 4 - 5 | 442 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |