Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105779.1 | NP_001099249.1 | 2 - 3 | 135 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |