Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008507.1 | NP_001008507.1 | 2 - 3 | 217 | 95 | |
XM_006246472.2 | XP_006246534.1 | 3 - 4 | 425 | 95 | ||
XM_006246473.2 | XP_006246535.1 | 2 - 3 | 1086 | 95 | ||
XM_006246474.2 | XP_006246536.1 | 2 - 3 | 554 | 95 | ||
XM_006246475.2 | XP_006246537.1 | 2 - 3 | 330 | 95 | ||
XM_008767786.1 | XP_008766008.1 | 2 - 3 | 413 | 95 | ||
GenBank mRNA | BC086453.1 | - | 2 - 3 | 217 | 7305216 | 95 |
EF445633.1 | - | 3 - 4 | 329 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |