Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031010.2 | NP_112272.2 | 9 - 10 | 1259 | 128 | |
XM_006246823.2 | XP_006246885.1 | 11 - 12 | 1540 | 128 | ||
XM_006246824.2 | XP_006246886.1 | 10 - 11 | 1435 | 128 | ||
GenBank mRNA | FQ233819.1 | - | 3 - 4 | 387 | 128 | |
L06040.1 | - | 9 - 10 | 1272 | 128 | ||
S69383.1 | - | 9 - 10 | 1259 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |