Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100674.1 | NP_001094144.1 | 8 - 9 | 862 | 76 | |
GenBank mRNA | BC160839.1 | - | 8 - 9 | 732 | 7458293 | 76 |
FQ231052.1 | - | 2 - 3 | 192 | 76 |
Gene Symbol: LOC103689956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767884.1 | XP_008766106.1 | 8 - 9 | 916 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |