Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107049.1 | NP_001100519.1 | 7 - 8 | 1181 | 66 | |
XM_006247310.2 | XP_006247372.1 | 7 - 8 | 2035 | 66 | ||
GenBank mRNA | BC162060.1 | - | 7 - 8 | 1124 | 7115660 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |