Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191822.1 | NP_001178751.1 | 6 - 7 | 840 | 143 | |
XM_006245894.2 | XP_006245956.1 | 5 - 6 | 2420 | 143 | ||
XM_008767537.1 | XP_008765759.1 | 6 - 7 | 2594 | 143 | ||
GenBank mRNA | Y17258.1 | - | 3 - 4 | 224 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |