Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030989.3 | NP_112251.2 | 9 - 10 | 1230 | 68 | |
XM_006246594.2 | XP_006246656.1 | 10 - 11 | 1659 | 68 | ||
XM_006246595.2 | XP_006246657.1 | 9 - 10 | 2016 | 68 | ||
GenBank mRNA | AF209191.1 | - | 6 - 7 | 911 | 68 | |
AY009504.1 | - | 4 - 5 | 436 | 68 | ||
BC081788.1 | - | 9 - 10 | 1159 | 7193583 | 68 | |
BC098663.1 | - | 9 - 10 | 1188 | 7384467 | 68 | |
FQ232659.1 | - | 9 - 10 | 1218 | 68 | ||
U90328.1 | - | 8 - 9 | 1099 | 68 | ||
X13058.1 | - | 8 - 9 | 1122 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |