Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009626.1 | NP_001009626.1 | 4 - 5 | 427 | 79 | |
GenBank mRNA | AM071386.1 | - | 4 - 5 | 386 | 79 | |
BC088180.1 | - | 4 - 5 | 427 | 7364928 | 79 | |
FQ209412.1 | - | 4 - 5 | 466 | 79 | ||
FQ209491.1 | - | 4 - 5 | 453 | 79 | ||
FQ209740.1 | - | 4 - 5 | 462 | 79 | ||
FQ210182.1 | - | 4 - 5 | 453 | 79 | ||
FQ210311.1 | - | 4 - 5 | 453 | 79 | ||
FQ210756.1 | - | 4 - 5 | 469 | 79 | ||
FQ210813.1 | - | 4 - 5 | 453 | 79 | ||
FQ211328.1 | - | 4 - 5 | 426 | 79 | ||
FQ218445.1 | - | 4 - 5 | 467 | 79 | ||
FQ218597.1 | - | 4 - 5 | 449 | 79 | ||
FQ218599.1 | - | 4 - 5 | 453 | 79 | ||
FQ218637.1 | - | 4 - 5 | 453 | 79 | ||
FQ218785.1 | - | 4 - 5 | 469 | 79 | ||
FQ218859.1 | - | 4 - 5 | 426 | 79 | ||
FQ218865.1 | - | 4 - 5 | 453 | 79 | ||
FQ219138.1 | - | 4 - 5 | 518 | 79 | ||
FQ219328.1 | - | 4 - 5 | 453 | 79 | ||
FQ219380.1 | - | 4 - 5 | 454 | 79 | ||
FQ219438.1 | - | 4 - 5 | 452 | 79 | ||
FQ219481.1 | - | 4 - 5 | 466 | 79 | ||
FQ219580.1 | - | 4 - 5 | 467 | 79 | ||
FQ219601.1 | - | 4 - 5 | 466 | 79 | ||
FQ219674.1 | - | 4 - 5 | 453 | 79 | ||
X15551.1 | - | 4 - 5 | 282 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |