Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106048.1 | NP_001099518.1 | 1 - 2 | 282 | 114 | |
XM_006252262.1 | XP_006252324.1 | 2 - 3 | 1051 | 114 | ||
XM_006252264.2 | XP_006252326.1 | 2 - 3 | 1512 | 114 | ||
XM_008770826.1 | XP_008769048.1 | 1 - 2 | 2486 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |