Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034937.2 | NP_001030109.2 | 4 - 5 | 513 | 101 | |
XM_006252509.2 | XP_006252571.1 | 4 - 5 | 587 | 101 | ||
GenBank mRNA | FM053767.1 | - | 4 - 5 | 441 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |