Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013051.1 | NP_001013069.1 | 7 - 8 | 454 | 147 | |
GenBank mRNA | BC158724.1 | - | 7 - 8 | 454 | 9090831 | 147 |
FQ220550.1 | - | 7 - 8 | 489 | 147 | ||
FQ221379.1 | - | 7 - 8 | 480 | 147 | ||
FQ225182.1 | - | 7 - 8 | 572 | 147 | ||
FQ227975.1 | - | 7 - 8 | 572 | 147 | ||
FQ232174.1 | - | 7 - 8 | 572 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |