Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106051.3 | NP_001099521.1 | 6 - 7 | 546 | 84 | |
NM_001270865.1 | NP_001257794.1 | 6 - 7 | 628 | 84 | ||
NM_001270866.1 | NP_001257795.1 | 6 - 7 | 546 | 84 | ||
GenBank mRNA | BC158790.1 | - | 6 - 7 | 537 | 7308742 | 84 |
FQ213565.1 | - | 6 - 7 | 546 | 84 | ||
FQ216373.1 | - | 6 - 7 | 630 | 84 | ||
FQ218750.1 | - | 6 - 7 | 519 | 84 | ||
FQ225558.1 | - | 3 - 4 | 306 | 84 | ||
FQ229329.1 | - | 6 - 7 | 520 | 84 | ||
FQ234562.1 | - | 3 - 4 | 306 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |