Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006963.1 | NP_001006964.1 | 3 - 4 | 609 | 74 | |
GenBank mRNA | BC083735.1 | - | 3 - 4 | 609 | 7191589 | 74 |
FQ210227.1 | - | 3 - 4 | 639 | 74 | ||
FQ212925.1 | - | 3 - 4 | 566 | 74 | ||
FQ218718.1 | - | 3 - 4 | 608 | 74 | ||
FQ229426.1 | - | 3 - 4 | 608 | 74 | ||
FQ230065.1 | - | 3 - 4 | 581 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |