Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107275.1 | NP_001100745.1 | 6 - 7 | 972 | 122 | |
XM_006252270.2 | XP_006252332.1 | 6 - 7 | 997 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |