Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007656.1 | NP_001007657.1 | 6 - 7 | 733 | 106 | |
XM_006239796.2 | XP_006239858.1 | 5 - 6 | 616 | 106 | ||
XM_008764512.1 | XP_008762734.1 | 5 - 6 | 783 | 106 | ||
XM_008764513.1 | XP_008762735.1 | 5 - 6 | 738 | 106 | ||
GenBank mRNA | BC083589.1 | - | 6 - 7 | 733 | 7132546 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |