Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135899.1 | NP_001129371.1 | 1 - 2 | 262 | 107 | |
GenBank mRNA | BC127545.1 | - | 1 - 2 | 262 | 7321962 | 107 |
FQ219104.1 | - | 1 - 2 | 306 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |