Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127572.1 | NP_001121044.1 | 3 - 4 | 267 | 92 | |
GenBank mRNA | BC161949.1 | - | 3 - 4 | 267 | 9025530 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |