Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106736.2 | NP_001100206.2 | 6 - 7 | 644 | 134 | |
GenBank mRNA | BC168873.1 | - | 6 - 7 | 609 | 134 | |
FQ214685.1 | - | 2 - 3 | 162 | 134 | ||
FQ214789.1 | - | 2 - 3 | 134 | |||
FQ223884.1 | - | 2 - 3 | 162 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons |