Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239906.2 | 6 - 7 | 945 | 83 | ||
XM_006239907.2 | XP_006239969.1 | 6 - 7 | 935 | 83 | ||
XM_006239908.2 | XP_006239970.1 | 5 - 6 | 851 | 83 | ||
XM_008764573.1 | 6 - 7 | 918 | 83 | |||
GenBank mRNA | BC078882.1 | - | 4 - 5 | 623 | 7115603 | 83 |
J04791.1 | - | 6 - 7 | 739 | 83 | ||
M16982.1 | - | 6 - 7 | 1003 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |