Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhcgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012978.1 | NP_037110.1 | 3 - 4 | 359 | 89 | |
GenBank mRNA | M26199.1 | - | 3 - 4 | 359 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons |