Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034133.1 | NP_001029305.1 | 3 - 4 | 533 | 76 | |
GenBank mRNA | BC105756.1 | - | 3 - 4 | 533 | 5623547 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |