Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037791.2 | NP_001032880.2 | 11 - 12 | 1343 | 119 | |
XM_006239770.2 | XP_006239832.1 | 11 - 12 | 1351 | 119 | ||
GenBank mRNA | BC169117.1 | - | 12 - 13 | 1540 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |