Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203366.2 | NP_976242.2 | 1 - 2 | 136 | 103 | |
NM_203367.2 | NP_976243.2 | 1 - 2 | 136 | 103 | ||
NM_203368.2 | NP_976244.2 | 1 - 2 | 136 | 103 | ||
NM_203369.2 | NP_976245.2 | 2 - 3 | 217 | 103 | ||
XM_006254150.2 | XP_006254212.1 | 2 - 3 | 321 | 103 | ||
XM_006254151.2 | XP_006254213.1 | 2 - 3 | 374 | 103 | ||
XM_006254152.2 | XP_006254214.1 | 2 - 3 | 233 | 103 | ||
XM_006254154.2 | XP_006254216.1 | 2 - 3 | 321 | 103 | ||
XM_006254155.2 | XP_006254217.1 | 2 - 3 | 373 | 103 | ||
XM_008771704.1 | XP_008769926.1 | 2 - 3 | 374 | 103 | ||
XM_008771705.1 | XP_008769927.1 | 2 - 3 | 372 | 103 | ||
XM_008771707.1 | XP_008769929.1 | 2 - 3 | 373 | 103 | ||
XM_008771708.1 | XP_008769930.1 | 2 - 3 | 371 | 103 | ||
XM_008771709.1 | XP_008769931.1 | 2 - 3 | 372 | 103 | ||
XM_008771710.1 | XP_008769932.1 | 2 - 3 | 371 | 103 | ||
XM_008771711.1 | XP_008769933.1 | 2 - 3 | 372 | 103 | ||
GenBank mRNA | BC065308.1 | - | 2 - 3 | 214 | 5599817 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |