Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012527.2 | NP_036659.2 | 4 - 5 | 521 | 112 | |
XM_008771690.1 | XP_008769912.1 | 5 - 6 | 467 | 112 | ||
GenBank mRNA | M18088.1 | - | 2 - 3 | 105 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Ion Channel Rattus norvegicus Inventoried Probe spans exons |