Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135157.1 | NP_001128629.1 | 66 | 60 | ||
GenBank mRNA | DQ872905.1 | - | 2 - 3 | 66 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |