Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slu7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100550.1 | NP_001094020.1 | 9 - 10 | 993 | 67 | |
XM_008767640.1 | XP_008765862.1 | 8 - 9 | 1768 | 67 | ||
XM_008767641.1 | XP_008765863.1 | 9 - 10 | 1440 | 67 | ||
XM_008767642.1 | XP_008765864.1 | 9 - 10 | 1645 | 67 | ||
GenBank mRNA | BC062243.1 | - | 9 - 10 | 997 | 67 | |
BC158865.1 | - | 9 - 10 | 959 | 7938574 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |