Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107034.1 | NP_001100504.1 | 7 - 8 | 962 | 59 | |
XM_006247036.2 | XP_006247098.1 | 7 - 8 | 1060 | 59 | ||
XM_006247037.2 | XP_006247099.1 | 5 - 6 | 2982 | 59 | ||
XM_008768036.1 | XP_008766258.1 | 7 - 8 | 849 | 59 | ||
XM_008768037.1 | XP_008766259.1 | 4 - 5 | 1661 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |