Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107073.1 | NP_001100543.1 | 3 - 4 | 361 | 108 | |
XM_006247889.2 | XP_006247951.1 | 3 - 4 | 410 | 108 | ||
XM_008768477.1 | XP_008766699.1 | 3 - 4 | 410 | 108 | ||
GenBank mRNA | BC167031.1 | - | 3 - 4 | 343 | 7099085 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |