Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107073.1 | NP_001100543.1 | 4 - 5 | 425 | 98 | |
XM_006247889.2 | XP_006247951.1 | 4 - 5 | 474 | 98 | ||
XM_008768477.1 | XP_008766699.1 | 4 - 5 | 474 | 98 | ||
GenBank mRNA | BC167031.1 | - | 4 - 5 | 407 | 7099085 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |