Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012575.3 | NP_036707.3 | 9 - 10 | 2173 | 69 | |
XM_006247706.2 | XP_006247768.1 | 7 - 8 | 1892 | 69 | ||
XM_006247707.1 | XP_006247769.1 | 10 - 11 | 2334 | 69 | ||
XM_006247708.2 | XP_006247770.1 | 7 - 8 | 1789 | 69 | ||
XM_006247709.2 | XP_006247771.1 | 7 - 8 | 1789 | 69 | ||
GenBank mRNA | D13212.1 | - | 9 - 10 | 2173 | 69 | |
M91563.1 | - | 7 - 8 | 1792 | 69 | ||
U08259.1 | - | 10 - 11 | 2333 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |