Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105800.1 | NP_001099270.1 | 10 - 11 | 2203 | 69 | |
XM_006246661.2 | XP_006246723.1 | 9 - 10 | 1718 | 69 | ||
XM_006246662.2 | XP_006246724.1 | 9 - 10 | 1705 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |