Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gngt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135767.1 | NP_001129239.1 | 2 - 3 | 187 | 71 | |
XM_006247208.2 | XP_006247270.1 | 2 - 3 | 298 | 71 | ||
XM_006247209.1 | XP_006247271.1 | 2 - 3 | 96 | 71 | ||
XM_006247210.1 | XP_006247272.1 | 2 - 3 | 96 | 71 | ||
XM_008768115.1 | XP_008766337.1 | 1 - 2 | 243 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus 1700 Inventoried Probe spans exons |