Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108243.2 | NP_001101713.1 | 3 - 4 | 344 | 84 | |
GenBank mRNA | BC168975.1 | - | 3 - 4 | 344 | 8365927 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |