Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prame | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001055257.4 | XP_001055257.2 | 3 - 4 | 980 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |