Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239725.2 | XP_006239787.1 | 1 - 2 | 52 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |