Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009602.1 | NP_001009602.1 | 11 - 12 | 895 | 82 | |
GenBank mRNA | BC087143.1 | - | 11 - 12 | 895 | 7128051 | 82 |
FQ219976.1 | - | 7 - 8 | 533 | 82 | ||
FQ228754.1 | - | 11 - 12 | 950 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |