Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009602.1 | NP_001009602.1 | 9 - 10 | 744 | 87 | |
GenBank mRNA | BC087143.1 | - | 9 - 10 | 744 | 7128051 | 87 |
FQ219976.1 | - | 5 - 6 | 382 | 87 | ||
FQ228754.1 | - | 9 - 10 | 799 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |