Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fosl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012954.1 | NP_037086.1 | 2 - 3 | 356 | 56 | |
GenBank mRNA | AY622611.1 | - | 2 - 3 | 353 | 56 | |
U18913.1 | - | 2 - 3 | 356 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons |