Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108709.3 | NP_001102179.2 | 4 - 5 | 370 | 103 | |
NM_001163156.3 | NP_001156628.1 | 4 - 5 | 389 | 103 | ||
XM_006240047.2 | XP_006240109.1 | 3 - 4 | 351 | 103 | ||
XM_006240048.2 | XP_006240110.1 | 4 - 5 | 409 | 103 | ||
XM_008764647.1 | XP_008762869.1 | 3 - 4 | 351 | 103 | ||
XM_008764648.1 | XP_008762870.1 | 4 - 5 | 824 | 103 | ||
GenBank mRNA | BC168860.1 | - | 4 - 5 | 370 | 7121808 | 103 |
CB547717.1 | - | 4 - 5 | 388 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |