Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108712.2 | NP_001102182.2 | 5 - 6 | 708 | 108 | |
XM_006240120.2 | XP_006240182.1 | 5 - 6 | 726 | 108 | ||
GenBank mRNA | BC166982.1 | - | 5 - 6 | 635 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |