Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108016.2 | NP_001101486.2 | 8 - 9 | 996 | 99 | |
XM_006239985.2 | XP_006240047.1 | 7 - 8 | 870 | 99 | ||
GenBank mRNA | CB547161.1 | - | 1 - 2 | 120 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |