Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nampt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177928.3 | NP_808789.1 | 7 - 8 | 989 | 94 | |
GenBank mRNA | AB081730.1 | - | 7 - 8 | 964 | 94 | |
AY831728.1 | - | 7 - 8 | 962 | 94 | ||
BC085681.1 | - | 7 - 8 | 989 | 7192109 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |