Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212522.1 | NP_997687.1 | 5 - 6 | 714 | 76 | |
XM_006240244.2 | XP_006240306.1 | 5 - 6 | 826 | 76 | ||
XM_008764770.1 | XP_008762992.1 | 5 - 6 | 826 | 76 | ||
XM_008764771.1 | XP_008762993.1 | 6 - 7 | 938 | 76 | ||
XM_008764772.1 | XP_008762994.1 | 6 - 7 | 920 | 76 | ||
GenBank mRNA | AY238343.1 | - | 5 - 6 | 714 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |